Learn More
Grant Instruments™ Stainless Steel Lid with Ring Sets
Product Code. 11466427
Change view
Click to view available options
For Use With (Equipment):
JBA12, JBN12, SAP12
JBA18, JBN18, SAP18
JBA5, JBN5, SAP5
SAP34
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11466427 JBA12, JBN12, SAP12 Each
11476427 JBA18, JBN18, SAP18 Each
11456427 JBA5, JBN5, SAP5 Each
11486427 SAP34 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11466427 Supplier Grant Instruments™ Supplier No. LF14
Only null left
Add to basket
Add to basket
Accommodate tall vessels whilst reducing evaporation. Grant Instruments Stainless Steel Lid with Ring Sets are flat with a range of variable hole diameters.
  • Stainless steel construction
  • Flat lids contain 2, 4 or 6 ring sets.
  • Variable hole diameter
  • Accommodates taller vessels
TRUSTED_SUSTAINABILITY
Specifications
Description 4-Ring Sets
For Use With (Equipment) JBA12, JBN12, SAP12
Material Stainless Steel