Learn More
Amsta Rostfritt Stainless Steel Lid For 608/609-100
Product Code. 30285026
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30285026 None Each
1 options
Product Code. 30285026 Supplier Amsta Rostfritt Supplier No. 5010450
  • ONLINE EXCLUSIVE PRICE
    £96.92
    £81.98 / Pack of 1
    Save £14.94
    15% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest Quality Stainless Steel lid For 608/609-100 Amsta transport container. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel