Learn More
Amsta Rostfritt Stainless Steel Lid For 605/606-10, 605-12
Product Code. 30284913
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30284913 None Each
1 options
Product Code. 30284913 Supplier Amsta Rostfritt Supplier No. 5010240
  • ONLINE EXCLUSIVE PRICE
    £31.25
    £28.33 / Pack of 1
    Save £2.92
    9% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest Quality Stainless Steel lid For 605/606-10. 605-12 Amsta buckets. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel