missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Stainless Steel Lab Frame

OHAUS™ Stainless Steel Lab Frame provides secure support for holding glassware at elevated positions.

Brand:  OHAUS™ 30392326

Additional Details : Weight : 30.60000kg

 View more versions of this product

Product Code. 15702379

  • £1590.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
Specifications

Specifications

Lab Frame Kit
Clamp, Lab Frame, Steel, CLR-FRAMESX
48.00 in.
305.00 mm
72.00 in.
Stainless Steel
1,219.000 mm
12.00 in.
1219 x 305 x 1829 mm (H x L x W)
1,829.00 mm
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers