missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, High Nitrogen, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, High Nitrogen, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Zinc/Selenium/Arsenic/Zirconium/Lead/Tin/Titanium/Sulfur/Phosphorus/Cobalt/Aluminium/Nitrogen/Tungsten/Molybdenum/Carbon/Copper/Vanadium/Silicon/Niobium/Nickel/Manganese/Chromium/Magnesium/Tantalum/Boron/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony -200 ppm/Zinc -100 ppm/Selenium -50 ppm/Arsenic -30 ppm/Zirconium -30 ppm/Lead -20 ppm/Tin 0.0026 %/Titanium 0.004 %/Sulfur 0.007 %/Phosphorus 0.0103 %/Cobalt 0.015 %/Aluminium 0.034 %/Nitrogen 0.087 %/Tungsten 0.104 %/Molybdenum 0.24 %/Carbon 0.269 %/Copper 0.438 %/Vanadium 0.54 %/Silicon 0.93 %/Niobium 1.48 %/Nickel 5.1 %/Manganese 6.79 %/Chromium 19.46 %/Magnesium (<100) ppm/Tantalum (<100) ppm/Boron (<4) ppm/Iron (<64) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction