missing translation for 'onlineSavingsMsg'
Learn More

Lauda™ Stainless Steel Hanging Rack

Product Code. 10522962
Click to view available options
Diameter (Metric) Min. Opening:
18 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10522962

Brand: Lauda™ UE 035

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Order Info

TemperatureMetricRange

Specifications

Type Inset Tube Rack
Holds Tubes (14 to 18 mm)
No. of Positions 11
Temperature (Metric) Maximum 100°C
Diameter (Metric) Min. Opening 18 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.