Learn More
Memmert™ Stainless Steel Grid, Electropolished
Product Code. 17105164 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17105164 IPP410eco Each
17119714 HPP1400eco Each
17119784 HPP260eco Each
17120105 IPP110eco Each
17155324 IPP260eco Each
17159874 HPP750eco Each
17160125 IPP1400eco Each
17160145 IPP2200eco Each
17169734 HPP2200eco Each
17169824 HPP410eco Each
17180265 IPP750eco Each
17189684 HPP110eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 17105164 Supplier Memmert™ Supplier No. IPP410ECOE28891
Additional electropolished stainless steel grid
  • Material: Stainless Steel
  • Electropolished
Specifications
For Use With (Equipment) IPP410eco
Material Stainless Steel