missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Greek Ascoloy, AISI 418, S41800, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Greek Ascoloy, AISI 418, S41800, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Tin/Oxygen/Aluminium/Niobium/Phosphorus/Cobalt/Nitrogen/Copper/Vanadium/Carbon/Molybdenum/Manganese/Silicon/Nickel/Tungsten/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.004 %/Tin 0.005 %/Oxygen 0.0056 %/Aluminium 0.006 %/Niobium 0.01 %/Phosphorus 0.019 %/Cobalt 0.03 %/Nitrogen 0.0434 %/Copper 0.069 %/Vanadium 0.17 %/Carbon 0.18 %/Molybdenum 0.32 %/Manganese 0.35 %/Silicon 0.4 %/Nickel 1.94 %/Tungsten 3.52 %/Chromium 12.42 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction