missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Tubing
Description
- Smooth and clean bore
- Supplied in coils
- Supplied cleaned and passivated
Specifications
Specifications
| Type | Tubing |
| Includes | Smooth and clean bore |
| Diameter (Metric) Inner | 2.2 mm |
| Diameter (Metric) Outer | 3.1 mm |
| Overall Length (Metric) | 5 m |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?