missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel gauze, 40 mesh woven from 0.25mm (0.01in) dia wire, Type 304, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 013478.RU
This item is not returnable.
View return policy
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel gauze, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
30 x 60 cm | |
40 mesh |
Woven from 0.25mm (0.01 in.) dia. wire | |
MFCD00148499 | |
Insoluble in water. | |
Wire Cloth |
RUO – Research Use Only