Learn More
Stainless Steel gauze, 325 mesh woven from 0.036mm (0.0014in) dia wire, Type 316
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Stainless Steel gauze, Type 316 |
| Name Note | 325 mesh woven from 0.036mm (0.0014 in.) dia. wire |
| Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
| Quantity | 75 x 75 mm |
| Mesh Size | 325 Mesh |
| Odor | Odorless |
| Form | Wire Cloth |
| MDL Number | MFCD00148499 |
| Solubility Information | Insoluble in water. |
Safety and Handling
- Stainless Steel gauze, Type 316
- Danger
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Nickel
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.