Learn More
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 013476.RR
Code : NEW
Additional Details : Weight : 0.01000kg
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel gauze | |
Fe:Cr:Ni; 70:19:11 wt.% | |
10 x 10 cm | |
200 Mesh |
Open area: 36%; Width of opening: 0.003 in. | |
MFCD00148499 | |
Insoluble in water. | |
Wire Cloth |
RUO – Research Use Only