missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Stainless Steel Forceps Serrated Jaws

Product Code. 17455812 Shop All Nickel-Electro Products
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17455812

Brand: Nickel Electro™ 7112

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory stainless steel forceps with serrated jaws and blunt ends.

  • Laboratory forceps
  • Stainless steel
  • Serrated jaws
  • 130 mm
  • Blunt ends

Specifications

Tip Style Straight, Blunt
Length (English) 5 in.
Material Stainless Steel
Length (Metric) 130 mm
Product Type Forceps
Sterility Non-sterile
Style Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.