missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Foil, 0.2mm (0.008 in.) thick, Alfa Aesar™

Brand:  Alfa Aesar 042255.GH

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100 x 100mm

 View more versions of this product

Product Code. 11359917

  • £35.90 / Each


Estimated Shipment date
from Supplier 30-04-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel
Foil, 0.2mm (0.008 in.) thick
MFCD00148499
Type 304
Fe:Cr:Ni; 70:19:11 wt%

Specifications

Stainless Steel
100 x 100mm
Silver-colored foil
Foil, 0.2mm (0.008 in.) thick
MFCD00148499
Silver
Type 304
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Videos
SDS