Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Product Code. 15325862
Change view
Click to view available options
For Use With (Equipment):
1000mL Flask
100mL Flask
250mL Flask
25mL Flask
500mL Flask
50mL Flask
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15325862 1000mL Flask Each
15365852 100mL Flask Each
15385852 250mL Flask Each
15315852 25mL Flask Each
15305862 500mL Flask Each
15335852 50mL Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15325862 Supplier Grant Instruments™ Supplier No. SC1000
Only null left
Add to basket
Add to basket
Holds flasks in place when used with Grant Aqua Pro Shaking Baths
  • Spring clamp provides security while allowing easy flask insertion/removal
  • For use with OLS and LSB Model Aqua Pro Shaking Water Baths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1000mL Flask
Type Flask Clamp
Material Stainless Steel