missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 013787.A1

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11341319

  • £101.00 / 1kg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel flake, Type 316-L
MFCD00148499
Insoluble in water.
0.4 to 1.2 Micron thick
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
1 kg
−325 Mesh
Flake
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only