Learn More
BINDER™ Expansion Rack, Includes Calibration
Product Code. 16636483 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers VD 115, VDL 115
BINDER Chambers VD 23, VDL 23
BINDER Chambers VD 56, VDL 56
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16636483 BINDER Chambers VD 115, VDL 115 Each
16616483 BINDER Chambers VD 23, VDL 23 Each
16626483 BINDER Chambers VD 56, VDL 56 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16636483 Supplier BINDER™ Supplier No. 86120013
  • ONLINE EXCLUSIVE PRICE
    £810.00
    £794.00 / Pack of 1
    Save £16.00
    1% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
Use this stainless steel expansion rack instead of the standard aluminum expansion rack. Includes calibration.
  • Stainless steel
  • Accessory rack
Specifications
Description Expansion racks - stainless steel expansion rack instead of standard aluminum expansion rack, incl. calibration
For Use With (Equipment) BINDER Chambers VD 115, VDL 115
Type Rack
Includes Calibration
Material Stainless Steel