missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Evaporating Dish

Brand:  Bochem™ 8396

Additional Details : Weight : 0.57600kg

 View more versions of this product

Product Code. 10525124

  • £39.05 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel (18/10)
Evaporating Dish
335 x 245 x 66 mm
245 mm
Rectangular
1 Ea.
335 mm
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers