missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, Zeron 100, S32760, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Duplex, Zeron 100, S32760, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Niobium/Tin/Aluminium/Carbon/Phosphorus/Vanadium/Cobalt/Nitrogen/Silicon/Manganese/Copper/Tungsten/Molybdenum/Nickel/Chromium/Bismuth/Cadmium/Calcium/Lead/Selenium/Zinc/Zirconium/Tantalum/Antimony/Hydrogen/Magnesium/Titanium/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.002 %/Oxygen 0.003 %/Niobium 0.005 %/Tin 0.006 %/Aluminium 0.017 %/Carbon 0.017 %/Phosphorus 0.026 %/Vanadium 0.09 %/Cobalt 0.123 %/Nitrogen 0.22 %/Silicon 0.24 %/Manganese 0.52 %/Copper 0.55 %/Tungsten 0.56 %/Molybdenum 3.61 %/Nickel 7.1 %/Chromium 25.5 %/Bismuth (<0.001) %/Cadmium (<0.001) %/Calcium (<0.002) %/Lead (<0.003) %/Selenium (<0.004) %/Zinc (<0.005) %/Zirconium (<0.005) %/Tantalum (<0.045) %/Antimony (0.001) %/Hydrogen (0.001) %/Magnesium (0.002) %/Titanium (0.004) %/Arsenic (0.0045) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction