missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, ASTM A182 Grade F51, S31803, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, Duplex, ASTM A182 Grade F51, S31803, Ø44x20mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Phosphorus/Carbon/Cobalt/Tungsten/Vanadium/Copper/Nitrogen/Silicon/Manganese/Molybdenum/Nickel/Chromium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium 0.02 %/Phosphorus 0.023 %/Carbon 0.031 %/Cobalt 0.032 %/Tungsten 0.035 %/Vanadium 0.06 %/Copper 0.15 %/Nitrogen 0.185 %/Silicon 0.29 %/Manganese 1.53 %/Molybdenum 3.28 %/Nickel 5.76 %/Chromium 22.4 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction