missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, Alloy 2304, S32304, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Duplex, Alloy 2304, S32304, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Niobium/Aluminium/Tin/Tungsten/Carbon/Phosphorus/Vanadium/Cobalt/Nitrogen/Copper/Molybdenum/Silicon/Manganese/Nickel/Chromium/Calcium/Lead/Titanium/Tantalum/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0019 %/Oxygen 0.0032 %/Niobium 0.004 %/Aluminium 0.005 %/Tin 0.008 %/Tungsten 0.015 %/Carbon 0.025 %/Phosphorus 0.028 %/Vanadium 0.061 %/Cobalt 0.064 %/Nitrogen 0.123 %/Copper 0.21 %/Molybdenum 0.249 %/Silicon 0.39 %/Manganese 1.26 %/Nickel 4.19 %/Chromium 22.5 %/Calcium (0.0003) %/Lead (0.001) %/Titanium (0.002) %/Tantalum (0.004) %/Arsenic (0.006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction