missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, Alloy 2205, S32205, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Duplex, Alloy 2205, S32205, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Oxygen/Tin/Aluminium/Niobium/Carbon/Phosphorus/Tungsten/Vanadium/Cobalt/Nitrogen/Copper/Silicon/Manganese/Molybdenum/Nickel/Chromium/Bismuth/Cadmium/Magnesium/Selenium/Lead/Zinc/Zirconium/Antimony/Hydrogen/Boron/Calcium/Arsenic/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.003 %/Oxygen 0.004 %/Tin 0.006 %/Aluminium 0.007 %/Niobium 0.011 %/Carbon 0.018 %/Phosphorus 0.023 %/Tungsten 0.024 %/Vanadium 0.063 %/Cobalt 0.104 %/Nitrogen 0.17 %/Copper 0.208 %/Silicon 0.46 %/Manganese 1.18 %/Molybdenum 3.2 %/Nickel 5.57 %/Chromium 22.6 %/Bismuth (<0.0001) %/Cadmium (<0.0001) %/Magnesium (<0.001) %/Selenium (<0.001) %/Lead (<0.003) %/Zinc (<0.005) %/Zirconium (<0.005) %/Antimony (0.001) %/Hydrogen (0.001) %/Boron (0.002) %/Calcium (0.002) %/Arsenic (0.003) %/Tantalum (0.02) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction