missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Double Tray Racks for Upright Freezers, 278mm wide

Designed for applications where the samples are either not stored in boxes or have odd sizes. Tenak™ Double Tray Racks for Upright Freezers can be used with all freezer brands.

Brand:  Tenak™ TE29404

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 12487010

  • £180.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Consist of a rack with two drawers, lid, and handle on front
  • Available in two different widths
  • Manufactured from stainless steel
Specifications

Specifications

Lid
Yes
Odd Sized Samples
1 Pack
420 x 278 x 225mm
225 mm
Double Tray Rack
Stainless Steel
Upright Freezers
1 Pack
278 mm
420 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers