Learn More
Description
Use Thermo Scientific™ Disposable Resin to refill stainless steel ion exchangers. The mixed-bed resins have documented quality monitoring and reliably function due to an advanced regeneration service. Each regenerated batch is monitored for both consistent capacity and quality.
This service is only available in Germany.
Specifications
Specifications
| Description | Resin 2 x 22 liters disposable |
| Material | Stainless Steel |
| Purification Method | Ion Exchange |
| Resin Type | Mixed Bed |
| For Use With (Equipment) | Mixed bed deionizer DI 2800 (part number 02.2800) |
| Type | Disposable Resin |
This service is only available in Germany.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.