missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
dimForUseWithEquipment,NoofPositions
Requires
Description,Material,WarrantySpec
- Allows a variety of beakers or erlenmeyer flasks to be placed directly onto the shelf in water bath
- Minimises evaporation
- Maintains temperature stability
Ringed lid, concentric, tall vessels, water bath
Specifications
Specifications
| Description | Flat concentric ringed lid |
| For Use With (Equipment) | For 14 L Unstirred Baths |
| Holds | 4 x 105 mm |
| Material | Stainless Steel |
| Warranty | 3 Years |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?