missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Steel Rack for Cryogenic Freezers

With spring locks mounted on each shelf to secure cryoboxes. Tenak™ Comfort Steel Rack for Cryogenic Freezers can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate.

Brand:  Tenak™ TE24520

Additional Details : Weight : 2.05000kg

 View more versions of this product

Product Code. 15200834

  • £89.45 / Each
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

  • Available in a range of heights
  • Can be used with all freezer models
  • Sold without cryoboxes
  • Manufactured from stainless steel
Specifications

Specifications

Spring Lock
Boxes
14
1 Pack
140 mm
140 mm
Comfort Rack
Stainless Steel
Chest freezer
140 x 140 x 775mm
775 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers