Learn More
Nickel Electro™ Stainless Steel Flask Clip
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
Brand: Nickel Electro™ FC0025
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
- No sharp edges to cut or score glassware
- Can be autoclaved
- Plastic base
- Easy to assemble and remove Flask Clips from Perforated tray
Specifications
25 mL Conical Flask | |
Flask Clip | |
Stainless Steel |