Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2
Product Code. 16662242 Shop All Ohaus Products
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Unit Size:
Each
Product Code. 16662242
Brand: Ohaus™ 30390876
Only null left
Add to basket
Add to basket
Adjusted to Meet or Exceed OIML F2 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance No Certificate
Material 3003-H14 Aluminum
Mass 1 mg
Tolerance 0.060 mg
Includes Protective Case