Learn More
Memmert™ Stainless Steel Bottom Drip Tray for IN75, 15 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN75 Incubator
Unit Size:
Each
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | IN75 Incubator |
| Material | Stainless Steel |
| Type | Bottom Drip Tray |
| Height (Metric) | Rim: 15 mm |