Learn More
Memmert™ Stainless Steel Bottom Drip Tray for IF260plus, 15 mm
Product Code. 17596825 Shop All Memmert Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material unitSize
17596825 Stainless Steel Each
1 options
Product Code. 17596825 Supplier Memmert™ Supplier No. IF260plusB29722
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) IF260plus Incubator
Material Stainless Steel
Type Bottom Drip Tray
Height (Metric) Rim: 15 mm