missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Basket for use with Buerkle™ PowderProof

Product Code. 10142512 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric):
84 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10142512 84 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142512 Supplier Buerkle™ Supplier No. 53022050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Buerkle™ Stainless Steel Basket is a spare part for Buerkle™ PowderProof™ Sampler.

  • For Sample Bottles PowderProof™
TRUSTED_SUSTAINABILITY

Specifications

Product Type Basket for Sample Bottles
Material Stainless Steel (V2A)
Includes Holding ring
For Use With (Application) 500 mL Bottles
For Use With (Equipment) PowderProof Sampler
Diameter (Metric) 84 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.