Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 25.00000kg
Description
- Mesh bottom
- Two handles on each end
- Functional and modern design
- Available for a variety of capacities
Specifications
Basket with Handle | |
Transonic Analogous Ultrasonic Unit, TP 680 DH, TP 690 | |
Handle | |
Stainless Steel |