Learn More
Masterflex™ Stainless Steel Barbed Y-Connector
Description
- Barbed Y-Connector
- Stainless steel
- Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Specifications
Specifications
| Description | Barb Connection |
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Tubing Size | 5/32 in. |
| Outlet Port | 5/32 in. |
| Port Size | 5/32 in. |
| Sterilization Method | Autoclave, Ethylene Oxide or Gamma |
| Thread Size | 5/32 in. |
| Product Type | Barbed Y Connector |