missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 316H, S31609, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 316H, S31609, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Boron/Zinc/Calcium/Lead/Aluminium/Phosphorus/Nitrogen/Cobalt/Vanadium/Oxygen/Copper/Carbon/Silicon/Manganese/Molybdenum/Nickel/Chromium/Arsenic/Tin/Titanium/Zirconium/Niobium/Tantalum/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.0001 %/Boron 0.0006 %/Zinc 0.001 %/Calcium 0.0014 %/Lead 0.002 %/Aluminium 0.004 %/Phosphorus 0.004 %/Nitrogen 0.006 %/Cobalt 0.007 %/Vanadium 0.007 %/Oxygen 0.016 %/Copper 0.021 %/Carbon 0.16 %/Silicon 0.64 %/Manganese 1.71 %/Molybdenum 2.79 %/Nickel 12.9 %/Chromium 17 %/Arsenic (0.001) %/Tin (0.002) %/Titanium (0.002) %/Zirconium (0.003) %/Niobium (0.005) %/Tantalum (0.005) %/Tungsten (0.0119) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction