missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AMS 5898, AISI 420*, S42027, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AMS 5898, AISI 420*, S42027, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Phosphorus/Cobalt/Tungsten/Copper/Vanadium/Nickel/Carbon/Manganese/Nitrogen/Silicon/Molybdenum/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.005 %/Phosphorus 0.014 %/Cobalt 0.02 %/Tungsten 0.02 %/Copper 0.035 %/Vanadium 0.05 %/Nickel 0.16 %/Carbon 0.3 %/Manganese 0.36 %/Nitrogen 0.39 %/Silicon 0.54 %/Molybdenum 0.99 %/Chromium 15.3 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering