missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 440F Se, S44023, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 440F Se, S44023, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Titanium/Aluminium/Zirconium/Arsenic/Tantalum/Oxygen/Tin/Niobium/Phosphorus/Nitrogen/Cobalt/Tungsten/Vanadium/Copper/Selenium/Nickel/Silicon/Molybdenum/Manganese/Carbon/Chromium/Cerium/Calcium/Antimony/Lead/Magnesium/Zinc |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0006 %/Titanium 0.0015 %/Aluminium 0.0018 %/Zirconium 0.002 %/Arsenic 0.004 %/Tantalum 0.004 %/Oxygen 0.005 %/Tin 0.0056 %/Niobium 0.011 %/Phosphorus 0.019 %/Nitrogen 0.027 %/Cobalt 0.032 %/Tungsten 0.041 %/Vanadium 0.116 %/Copper 0.13 %/Selenium 0.17 %/Nickel 0.265 %/Silicon 0.49 %/Molybdenum 0.5 %/Manganese 0.95 %/Carbon 0.98 %/Chromium 17.01 %/Cerium (<0.005) %/Calcium (0.0003) %/Antimony (0.002) %/Lead (0.002) %/Magnesium (0.004) %/Zinc (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction