missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel, AISI 431, S43100, ARMI∣MBH

Product Code. 30315725
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30315725

Brand: ARMI/MBH MBH13X 43100 AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI/MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. Their offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbon∣Chromium∣Cobalt∣Copper∣Grade∣Manganese∣Molybdenum∣Nickel∣Niobium∣Nitrogen∣Phosphorus∣Silicon∣Tin∣Tungsten∣Vanadium
Quantity 50 g
Weight 50 g
Form Pieces
Concentration or Composition (by Analyte or Components) Tin 0.004 %∣Tungsten 0.004 %∣Niobium 0.006 %∣Phosphorus 0.0199 %∣Cobalt 0.0239 %∣Vanadium 0.0577 %∣Nitrogen 0.075 %∣Molybdenum 0.0768 %∣Copper 0.134 %∣Carbon 0.166 %∣Manganese 0.378 %∣Silicon 0.535 %∣Nickel 2.1 %∣Chromium 16.39 %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.