missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 420, S42000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, AISI 420, S42000, 100g
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Tin/Aluminium/Tungsten/Phosphorus/Cobalt/Nitrogen/Copper/Vanadium/Molybdenum/Nickel/Silicon/Carbon/Manganese/Chromium/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Titanium 0.002 %/Niobium 0.004 %/Tin 0.004 %/Aluminium 0.008 %/Tungsten 0.011 %/Phosphorus 0.018 %/Cobalt 0.021 %/Nitrogen 0.0294 %/Copper 0.044 %/Vanadium 0.045 %/Molybdenum 0.052 %/Nickel 0.19 %/Silicon 0.35 %/Carbon 0.39 %/Manganese 0.47 %/Chromium 12.82 %/Oxygen [0.0068] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction