missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 314, S31400, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, AISI 314, S31400, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Calcium/Tungsten/Niobium/Aluminium/Phosphorus/Nitrogen/Carbon/Vanadium/Cobalt/Copper/Molybdenum/Manganese/Nickel/Silicon/Chromium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Calcium (0.0025) %/Tungsten (0.013) %/Niobium (0.02) %/Aluminium (0.021) %/Phosphorus (0.025) %/Nitrogen (0.029) %/Carbon (0.033) %/Vanadium (0.095) %/Cobalt (0.133) %/Copper (0.21) %/Molybdenum (0.24) %/Manganese (1.6) %/Nickel (18.8) %/Silicon (2.28) %/Chromium (24.3) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction