missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 309, S30900, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 309, S30900, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Arsenic/Tin/Niobium/Phosphorus/Tungsten/Carbon/Vanadium/Cobalt/Silicon/Molybdenum/Copper/Manganese/Nickel/Chromium/Boron/Magnesium/Sulfur/Zirconium/Oxygen/Titanium/Aluminium/Nitrogen/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0018 %/Arsenic 0.0065 %/Tin 0.012 %/Niobium 0.021 %/Phosphorus 0.029 %/Tungsten 0.063 %/Carbon 0.066 %/Vanadium 0.073 %/Cobalt 0.248 %/Silicon 0.3 %/Molybdenum 0.357 %/Copper 0.43 %/Manganese 1.61 %/Nickel 12.2 %/Chromium 22.42 %/Boron (<0.001) %/Magnesium (<0.001) %/Sulfur (0.002) %/Zirconium (0.002) %/Oxygen (0.0029) %/Titanium (0.003) %/Aluminium (0.01) %/Nitrogen (0.058) %/Iron (62) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction