missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 304L, S30403, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, AISI 304L, S30403, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Aluminium/Titanium/Oxygen/Arsenic/Niobium/Tin/Carbon/Phosphorus/Tungsten/Vanadium/Nitrogen/Cobalt/Silicon/Molybdenum/Copper/Manganese/Nickel/Chromium/Lead/Magnesium/Selenium/Cerium/Tantalum/Zinc/Zirconium/Hydrogen/Antimony/Calcium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0012 %/Aluminium 0.003 %/Titanium 0.0056 %/Oxygen 0.006 %/Arsenic 0.007 %/Niobium 0.013 %/Tin 0.013 %/Carbon 0.024 %/Phosphorus 0.034 %/Tungsten 0.056 %/Vanadium 0.076 %/Nitrogen 0.081 %/Cobalt 0.156 %/Silicon 0.43 %/Molybdenum 0.462 %/Copper 0.54 %/Manganese 1.39 %/Nickel 8.17 %/Chromium 18.34 %/Lead (<0.001) %/Magnesium (<0.001) %/Selenium (<0.001) %/Cerium (<0.002) %/Tantalum (<0.005) %/Zinc (<0.005) %/Zirconium (<0.005) %/Hydrogen (0.0008) %/Antimony (0.002) %/Calcium (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction