missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 3 Model 300W Cooled Incubator, 290 L, Stainless Steel

Product Code. 11857592
Click to view available options
Capacity (Metric):
290 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11857592

Brand: LMS™ 300

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Temperature Range: -10°C to +50°C

  • Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
  • Reliable and accurate temperature control
  • Standard single and dual temperature, plus programmable models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests

Specifications

Certifications/Compliance ISO 9001:2008
Controller Type Standard digital
Description Freestanding incubator
Capacity (Metric) 290 L
Chamber Material Stainless Steel
Convection Type Fan-assisted Circulation
Dimensions (D x W) Shelf 510 mm x 390 mm
Lighting Optional Fluorescent/UV
Material Stove Enameled Mild Steel
No. of Shelves 3
Temperature Range (Metric) -10°C to +50°C
Type Refrigerated Incubator
Display Digital
Insulation Foamed polyurethane
Model 300W
No. of Doors 1
Refrigerant CFC-free
Shelf Area (Metric) 510 x 390 mm
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Warranty 2 Years
Width (English) 26.8 in.
Width (English) Interior 21.6 in.
Width (Metric) 680 mm
Width (Metric) Interior 550 mm
Height (English) 71.2 in.
Height (English) Interior 46.1 in.
Height (Metric) 1810 mm
Height (Metric) Interior 1170 mm
Depth (English) Interior 17.7 in.
Depth (Metric) Interior 450 mm
Weight (Metric) 130 kg
Electrical Requirements 240 V, 50 Hz
Voltage 230 V
Frequency 50 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.