Learn More
Julabo™ Stainless Steel 2-Adapter
Click to view available options
Connection Type:
M30 x 1.5 Female to M16 x 1 Male
M30 x 1.5 Male to M16 x 1 Male
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45 Temperature Control System |
| Type | 2-Adapters |
| Material | Stainless Steel |
| Connection Type | M30 x 1.5 Male to M16 x 1 Male |