missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Multiple Dispensing Manifold

Conveniently distribute to or aspirate from multi-well plates. Socorex™ Multiple Dispensing Manifold fits Dosys™ syringes as well as the Acura™ 865 microdispenser.

Brand:  Socorex™ 1.170.054

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11720627

  • £114.00 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel construction.
  • PTFE construction stoppers.
  • Allows distribution or aspiration with 24- and 96-well plates.
  • Tightly fits onto Dosys™ syringes and the Acura™ microdispenser.
  • Fully autoclavable at 121°C / 250°F.
Specifications

Specifications

Dispensing Manifold
4
Dosys™ syringes, Acura™ microdispenser
1/pk.
Autoclavable
Stainless Steel, PTFE
PTFE Stoppers
121°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers