Learn More
Thermo Scientific™ Stained Brucella Suspensions
Description
Rapidly detect and quantify specific antibodies to Brucella abortus and Brucella melitensis in sera with this stained suspension.
Rapidly detect and quantify specific antibodies to Brucella abortus and Brucella melitensis in sera with Thermo Scientific™ Remel™ Stained Brucella Suspensions. Stained Brucella Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.
Use Stained Brucella Suspensions for the qualitative detection of Brucella abortus and Brucella melitensis antibodies in sera in the investigation of pyrexia and in standard Widal tests1, 2, 3.
- Identifies major groups of Brucella serotypes
- Provides accurate, reliable results
- Simple, easy-to-use tests
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Stained Brucella Suspensions |
| Detectable Analytes | Brucella abortus Antibodies |
| Includes | Stained Brucella Suspensions |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.