Learn More
Agilent BioTek Stage Insert CellASIC ONIX2 XT
Product Code. 16572162 Shop All Agilent BioTek Products
Click to view available options
:
Stage Insert CellASIC ONIX2 XT
Product Code. 16572162
Brand: Agilent BioTek 1530007BT
Stage Insert CellASIC ONIX2 XT is for use with MilliporeSigma heated ONIX2 manifold
  • Includes plate stage and holder, tubing bracket
  • Plate sold separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Agilent BioTek Microplate Washer and Reader Instruments