missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540130
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STAG2 is a component of the cohesin complex. This complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.Specifications
STAG2 | |
Polyclonal | |
Unconjugated | |
STAG2 | |
9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q8N3U4 | |
STAG2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human STAG2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |