Learn More
BINDER™ Stacking Frame
Product Code. 16674182 Shop All BINDER Products
Change view
Click to view available options
Height (Metric):
1955 mm
Unit Size:
Each
Product Code. Height (Metric) Unit Size
16674182 1955 mm Each
1 options
Product Code. 16674182 Supplier BINDER™ Supplier No. 90510040
  • ONLINE EXCLUSIVE PRICE
    £1510.00
    £1409.00 / Pack of 1
    Save £101.00
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This stable, vibration-free Stacking Frame is designed for the safe stacking of two BINDER™ CB 170 CO2 Incubators. on casters with stop brake for safe stacking of two CB series units
  • Stable, vibration-free stack on stacking frame
  • Designed for the safe stacking of two BINDER™ CB 170 CO2 Incubators
  • Features casters with stop brake
Specifications
Description Stacking frame - stable, vibration-free stacking frame on casters with stop brake for safe stacking of two CB series units
For Use With (Equipment) BINDER Chambers CB/CBF/CB-S 170
Type Stacking Frame
Includes Casters
Height (Metric) 1955 mm