Learn More
Gilac™ Stackable Reinforced Crate with Handles
Product Code. p-200108419
Click to view available options
Color:
Gray
White
Unit Size:
Each
Product Code. 30245678
Brand: Gilac™ G154620
Only null left
Add to basket
Add to basket
Reinforced storage container in food-grade plastic (HDPE). Stackable bin with multi-purpose handles, ideal for handling.
  • Reinforced 545 x 390 mm storage container in food-grade plastic (HDPE)
  • Stackable bin with multi-purpose handles, ideal for handling. Ideal for negative or positive cold storage
  • This reinforced, professional-quality bin is designed for intensive use
Specifications
Product Type Storage Container
Color Gray