Learn More
Manutan Stack and nest storage bin, blue, NXT4, 7.5 litre
Product Code. 12841999
Encompass_Preferred
Click to view available options
Color:
Blue
Unit Size:
Each
Product Code. 12841999
Brand: Manutan A344358
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The ultimate space-saving storage containers that stack and nest. Resistant to most chemicals, including oil and acids. Temperature range from -15°C to +50°C (dependant on use). Manufactured from polypropylene. Lifting handle on larger bins. Eco-white - non-pigmented colour, offering the same features and quality as red and blue for less money. Supplied with labels. Optional dividers run back to front, or left to right dependent on bin size.
  • Type: A4
  • Blue
  • Dimensions (H x W x D): 130mm x 240mm x 355mm
Specifications
Color Blue